Learn More
Honeywell™ Clearflow 3 Compressed Airline Filtration Unit, Portable Stand Model
Delivers high-quality breathable air to a range of respiratory devices.
Brand:  Honeywell™ 1763904
Product Code. 15816435
  • 1375.00 € / 1 pair
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Floor model on portable stand with 2 CEJN outlets
  • Supplies breathable air for up to 4 workers using supplied respirators, or 2 operators and auxiliary air-driven tools
  • Uses coalescing filter and carbon absorber filter to remove oil vapor and odor
  • High filtration capacity due to high flow system, high efficiency of coalescing filter, and high capacity of absorber carbon filter
  • Easy use and easy maintenance due to new one-piece compact design
  • Provides security to worker via accurate and reliable differential pressure indicator
  • Filter
    - Coalescing filter : 1000 hours with a filtration at 99,97% to 0.01 micron - Charcoal filter : 1000 hours (or when odors are detected inside respiratory device) Filters are compliant with ISO 8573 -1 Compressed Air - Part 1: Pollutants and purity classes) which specifies the quality class of compressed air for the presence of particles, water or oil, irrespective of the mode of generation of the compressed air)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Breathable air/air for air-driven tools
Respirators/air-driven tools
8 bar
1600 L/min.
Aluminum body filters, plastic and brass regulators, polycarbonate bowl, nitrile seals, ABS cover
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Honeywell™ Clearflow 3 Compressed Airline Filtration Unit, Portable Stand Model > Airflow: 1600L/mn