missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens IPA VH1 Sterile Disinfectant
BIOPHARMA

Ready to use sterile surface disinfectant based on IPA 70%

Brand:  Diversey™ 101104901


 View more versions of this product

Product Code. 17633235

  • 353.00 € / 20 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ClearKlens IPA is a ready to use disinfectant for the Life Sciences cleanroom industry. ClearKlens IPA is based on 70% isopropyl alcohol. ClearKlens IPA is available as 900 mL bag in bottle trigger spray, and as 1 L bottle or 5 L canister. The 900 mL bottle has been filtered to 0.2 μm and irradiated. The 1 L/5 L has been filtered to 0.2 μm and aseptically filled.

Key properties

  • ClearKlens IPA is a solution of 70% isopropyl alcohol and 30% water for injection (USP quality).
  • ClearKlens IPA is a ready to use product for the disinfection of work surfaces and process equipment in cleanrooms
  • ClearKlens IPA 900 mL trigger spray is an antisuck back- bag in bottle delivery system to eliminate product contamination during use.
  • BPR approved pack and formulation
  • Double bagged for easy transfer into the cleanroom.

Benefits

  • Ready to use
  • 0.2 μm filtered
  • Gamma irradiated/aseptically filled and double bagged
  • Detex labelled for proof of irradiation
  • Double bagged for easy transfer into the cleanroom
  • Odorless and Non tainting
  • Broad spectrum of activity
Specifications

Specifications

Surface Disinfectant
Sterile
For ES, GB, IE, IT, PT only
5 L
6 to 9
70% IPA/30% WFI USP
4 Pk.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ClearKlens IPA VH1

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Propan-2-ol

SDS
Documents

Documents

Certificats
Promotions

Promotions

Each lot is provided with: Certificate of Analysis, Certificate of Sterility, Certificate of Irradiation