Learn More
Epredia™ ClearVue™ Coverslipper Coverslip Hoppers
Designed for use with the Epredia™ ClearVue™ coverlipper. ClearVue™ hoppers streamline coverslipping workflow.
Brand:  Epredia™ A79210051
95.76 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 12636376
  • 117.00 € / Pack of 2
Explore more special offers
Description
Description
  • Makes integration into the laboratory effortless
  • Ships preloaded with 500 or 575 slips each
  • Inserts quickly into the Epredia™ ClearVue™ coverslipper without workflow interruption
  • Requires no manual handling of glass
  • Dispenses a clean coverslip each and every time
Specifications
Specifications
Coverslip Hopper
2 x 500 Hoppers/Coverslips
0.16 to 0.19 mm (# 1.5)
Epredia ClearVue Coverslipper
24 mm
50 mm