missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CO2 Independent Medium
GREENER_CHOICE

A non-HEPES proprietary medium used for supporting cell growth for a variety of suspension and adherent mammalian cells.

Brand:  Gibco™ 18045070

 View more versions of this product

Product Code. 11590536

  • 584.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • CO2 Independent Medium is a non-HEPES proprietary medium used for supporting cell growth for a variety of suspension and adherent mammalian cells such as epithelial, fibroblast, and lymphoid cell lines without a CO2 incubator
  • CO2 Independent Medium is ideal for transporting cells or tissue, and for the handling of mouse embryos under atmospheric conditions
  • Gibco™ CO2 Independent Medium contains a unique buffering system composed of mono and dibasic sodium phosphate and β-glycerophosphate
  • A small amount of sodium bicarbonate has been included in the formulation to meet essential bicarbonate dependent functions
  • No synthetic buffers are utilized, thus eliminating any cytotoxic effects associated with such buffering systems
  • Additionally, Gibco™ CO2 Independent Medium has been formulated with components that enhance cellular production and utilization of CO2
  • Store at 2°C to 8°C, protected from light

Modified as follows

  • With: phenol red, sodium pyruvate
  • Without: L-glutamine, HEPES

Using CO2 Independent Medium

  • CO2 Independent Medium contains no lipids or growth factors therefore it requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • The CO2 Independent Medium does not require a CO2 environment to maintain physiological pH
  • For maximum growth performance some cell lines may require either direct or sequential adaptation to CO2 Independent Medium
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 13485 standards, FDA
1 X
CO2 Independent Medium
No Glutamine, No HEPES
10 x 500 mL
2°C to 8°C
Liquid
Phenol Red, Sodium Pyruvate
Mammalian cell culture
For research use only
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ CO2 Independent Medium > 10 x 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.