missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CO2 Independent Medium

A non-HEPES proprietary medium used for supporting cell growth for a variety of suspension and adherent mammalian cells.
77.25 € - 584.00 €
Specifications
Concentration | 1 X |
---|---|
Form | Liquid |
Product Type | CO2 Independent Medium |
With Additives | Phenol Red, Sodium Pyruvate |
Without Additives | No Glutamine, No HEPES |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15420604
![]() |
Gibco™
18045088 |
500 mL |
106.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11580536
![]() |
Gibco™
18045054 |
500 mL |
77.25 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11590536
![]() |
Gibco™
18045070 |
10 x 500 mL |
584.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- CO2 Independent Medium is a non-HEPES proprietary medium used for supporting cell growth for a variety of suspension and adherent mammalian cells such as epithelial, fibroblast, and lymphoid cell lines without a CO2 incubator
- CO2 Independent Medium is ideal for transporting cells or tissue, and for the handling of mouse embryos under atmospheric conditions
- Gibco™ CO2 Independent Medium contains a unique buffering system composed of mono and dibasic sodium phosphate and β-glycerophosphate
- A small amount of sodium bicarbonate has been included in the formulation to meet essential bicarbonate dependent functions
- No synthetic buffers are utilized, thus eliminating any cytotoxic effects associated with such buffering systems
- Additionally, Gibco™ CO2 Independent Medium has been formulated with components that enhance cellular production and utilization of CO2
- Store at 2°C to 8°C, protected from light
Modified as follows
- With: phenol red, sodium pyruvate
- Without: L-glutamine, HEPES
Using CO2 Independent Medium
- CO2 Independent Medium contains no lipids or growth factors therefore it requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
- The CO2 Independent Medium does not require a CO2 environment to maintain physiological pH
- For maximum growth performance some cell lines may require either direct or sequential adaptation to CO2 Independent Medium
Specifications
1 X | |
CO2 Independent Medium | |
No Glutamine, No HEPES |
Liquid | |
Phenol Red, Sodium Pyruvate |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ CO2 Independent Medium