Learn More
Cobalt(II,III) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
130.65 € - 557.78 €
Chemical Identifiers
CAS | 1308-06-1 |
---|---|
Molecular Formula | Co3O4 |
Molecular Weight (g/mol) | 240.80 |
MDL Number | MFCD00010939 |
InChI Key | UBEWDCMIDFGDOO-UHFFFAOYSA-N |
Synonym | oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt |
PubChem CID | 11651651 |
IUPAC Name | oxocobalt;oxo(oxocobaltiooxy)cobalt |
SMILES | [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11300959
|
Thermo Scientific Alfa Aesar
010695.09 |
10 g |
130.65 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11390949
|
Thermo Scientific Alfa Aesar
010695.18 |
50 g |
557.78 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in artificial photosynthesis. It is used as a magnetic semiconductor. It is also employed in supercapacitors and lithium-ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1308-06-1 | |
240.80 | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
11651651 | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Co3O4 | |
MFCD00010939 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt | |
oxocobalt;oxo(oxocobaltiooxy)cobalt |
Specifications
900°C (decomposition) | |
Black | |
Co3O4 | |
UN3077 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
oxocobalt;oxo(oxocobaltiooxy)cobalt | |
11651651 | |
99.9985% | |
6.11 g/mL | |
Cobalt(II,III) oxide |
1308-06-1 | |
Powder | |
MFCD00010939 | |
14,2429 | |
Insoluble in water.Soluble in acids and alkalies. | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] | |
240.80 | |
240.8 | |
Puratronic™ | |
(metals basis) |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
H334-H350i
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 215-157-2
missing translation for 'rtecsNumber' : GG2500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Cobalt(II,III) oxide
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350i-May cause cancer by inhalation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Cobalt oxide
RUO – Research Use Only