missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate heptahydrate, Honeywell™

Reagent Grade,≥99%

75.27 € - 552.00 €

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
PubChem CID 61444
IUPAC Name cobalt(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15607770
View Documents
Honeywell Research Chemicals
C6768-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15637770
View Documents
Honeywell Research Chemicals
C6768-250G
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15617770
View Documents
Honeywell Research Chemicals
C6768-1KG
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15627770
View Documents
Honeywell Research Chemicals
C6768-2.5KG
2.5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

10026-24-1
281.09
MEYVLGVRTYSQHI-UHFFFAOYSA-L
61444
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
CoH14O11S
MFCD00149658
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
cobalt(2+);sulfate;heptahydrate

Specifications

10026-24-1
CoSO4 · 7H2O
UN3077
MEYVLGVRTYSQHI-UHFFFAOYSA-L
cobalt(2+);sulfate;heptahydrate
61444
≥99%
Cobalt(II) sulfate heptahydrate
CoH14O11S
MFCD00149658
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
281.09
281.1g/mol
2.03g/mL at 25°C
Videos
Safety and Handling

Safety and Handling

P201-P261-P280-P284-P304 + P340-P308 + P313

missing translation for 'rtecsNumber' : GG3200000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.