Learn More
Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
210.00 € - 625.00 €
Chemical Identifiers
CAS | 10026-24-1 |
---|---|
Molecular Formula | CoH14O11S |
Molecular Weight (g/mol) | 281.09 |
MDL Number | MFCD00149658 |
InChI Key | MEYVLGVRTYSQHI-UHFFFAOYSA-L |
Synonym | cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o |
PubChem CID | 61444 |
SMILES | O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11330959
|
Thermo Scientific Alfa Aesar
010696.14 |
25 g |
210.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320959
|
Thermo Scientific Alfa Aesar
010696.22 |
100 g |
625.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) sulfate heptahydrate is used in the preparation of pigments and other cobalt salts. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10026-24-1 | |
281.09 | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
61444 |
CoH14O11S | |
MFCD00149658 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Specifications
10026-24-1 | |
4 | |
CoH14O11S | |
MFCD00149658 | |
14,2448 | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
cobalt(2+);sulfate;heptahydrate | |
61444 | |
99.999% | |
Odorless | |
(metals basis) |
Red | |
Crystalline Aggregates | |
CoSO4·7H2O | |
UN3077 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O | |
281.09 | |
281.10 (154.99 Anhydrous) | |
Puratronic™ | |
2.03g/mL | |
Cobalt(II) sulfate heptahydrate |
Safety and Handling
GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'rtecsNumber' : GG3200000
missing translation for 'tsca' : TSCA
- Cobalt(II) sulfate heptahydrate
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1A
- Reproductive toxicity Category 1A
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Acute toxicity Category 4
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only