missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ PCR-free ES DNA Library Prep Kits for Illumina Systems

The Invitrogen Collibri PCR-free ES DNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-genome sequencing (WGS) applications and is suitable for all genomes.

630.00 € - 3060.00 €

Specifications

Format Kit
Sequencing Type Genome and DNA Sequencing
For Use With (Application) Whole Genome Sequencing, Resequencing, De Novo Assembly
Libraries Fragment Library
Product Line Collibri™
View More Specs

Products 9
Product Code Brand Product Type No. of Reactions Price Quantity & Availability  
Product Code Brand Product Type No. of Reactions Price Quantity & Availability  
17767443
View Documents Promotion Details
Invitrogen™
A38545024W
PCR-free ES DNA Library Prep Core Kit 24 Preps
630.00 €
Pack of 24
Please to purchase this item. Need a web account? Register with us today!
17777443
View Documents Promotion Details
Invitrogen™
A38603096W
PCR-free ES DNA Library Prep Core Kit 96 Preps
2315.00 €
Pack of 96
Please to purchase this item. Need a web account? Register with us today!
16544301
View Documents Promotion Details
Invitrogen™
A38545024
PCR-free ES DNA Library Prep Kit with CD Indexes 24 Preps
814.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
16564301
View Documents Promotion Details
Invitrogen™
A38603096
PCR-free ES DNA Library Prep Kit with CD Indexes 96 Preps
2950.00 €
96 reactions
Please to purchase this item. Need a web account? Register with us today!
16554301
View Documents Promotion Details
Invitrogen™
A38602024
PCR-free ES DNA Library Prep Kit with UD Indexes (Set A, 1-24) 24 Preps
902.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
16594311
View Documents Promotion Details
Invitrogen™
A43602024
PCR-free ES DNA Library Prep Kit with UD Indexes (Set B, 25-48) 24 Preps
908.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
16504321
View Documents Promotion Details
Invitrogen™
A43603024
PCR-free ES DNA Library Prep Kit with UD Indexes (Set C, 49-72) 24 Preps
916.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
16514321
View Documents Promotion Details
Invitrogen™
A43604024
PCR-free ES DNA Library Prep Kit with UD Indexes (Set D, 73-96) 24 Preps
916.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
17276523
View Documents Promotion Details
Invitrogen™
A38608196
PCR-free ES DNA Library Prep Kit with UD Indexes (Set A-D, 1-96) 96 Preps
3060.00 €
96 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Collibri PCR-free ES DNA Library Preparation Kit is designed for fast and convenient library preparation from inputs as low as 100 ng intact genomic DNA or already prepared amplicons (>300 bp) of any species. The entire library preparation protocol takes about two hours for the PCR-free workflow. If needed, Collibri Library Amplification Master Mix with primers can be bought separately. Or see the Collibri ES DNA Library Preparation Kit, designed for fast and convenient library preparation with PCR from a wide range of inputs (1–500 ng).

The Collibri PCR-free ES DNA Library Preparation Kit is designed for fast and convenient library preparation from inputs as low as 100 ng intact genomic DNA or already prepared amplicons (>300 bp) of any species. The entire library preparation protocol takes about two hours for the PCR-free workflow. If needed, Collibri Library Amplification Master Mix with primers can be bought separately. Or see the Collibri ES DNA Library Preparation Kit, designed for fast and convenient library preparation with PCR from a wide range of inputs (1–500 ng).

The Collibri PCR-free ES DNA Library Preparation Kit:
• Includes all necessary components in a single kit* for quick use
• Is empowered on miniaturization liquid handlers to cut on costs
• Provides in-process visual feedback for easier handling
•Is suitable for conventional automation to increase throughput

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules, except for the Core kit, which enables Collibri combination with desired indexed adapters

The Collibri PCR-free ES DNA Library Preparation Kit contains inert dyes that provide visual feedback of preparation progress: the reaction mixes change color in response to the addition of critical components at each step. Moreover, the enzymatic shearing (ES) module of the kit generates a consistently high yield of library, which is enough for library quantification  and sequencing and multiple runs on Illumina instruments.

Applications
The Collibri PCR-free ES DNA Library Preparation Kit is recommended for:
Whole genome sequencing
• Resequencing
• De novo assembly

Specifications

Specifications

Kit
Whole Genome Sequencing, Resequencing, De Novo Assembly
Collibri™
Genome and DNA Sequencing
Fragment Library
Library Generation
Videos
SDS
Documents

Documents

Product Certifications