missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems

Kits are designed for next-generation sequencing (NGS) on Illumina systems and support whole-transcriptome sequencing (human, mouse, rat), and mRNA sequencing (compatible with all species) applications.

Brand:  Invitrogen™ A39003024

1636.20 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 16305155

  • 2045.00 € / Pack of 24

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• Collibri Stranded RNA Library Prep Kit (24 preps). Store at -20°C.
• Collibri Library Cleanup Kit (24 preps). Store at 4°C.
• Collibri H/M/R rRNA Depletion Kit (24 preps). Store at 4°C.

Kit
Total RNA
Sequencing
24 Reactions
Collibri™, Illumina™
24 preps
Dry Ice
The product contains modules A39117024, A39118024, A39115024, A39100024
Human, Mouse, Rat
HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 & 2000, NovaSeq 6000
RNA Prep, Depletion Kit
cDNA Libraries
Stranded RNA Library Prep Kit
Transcriptome Sequencing
Adaptor Hybridization, Cleanup, Fragmentation, Indexing, Ligation, PCR Amplification, rRNA Depletion, Reverse Transcription
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems > Stranded RNA Library Prep Kit, and Human/Mouse/Rat rRNA Depletion Kit; 24 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.