Learn More
SCHOTT AG Lighting and Imaging™ Column Adapter for Combi Light Guide
Column Adapter for use with Schott Combi Light Guide 170 402.
Brand:  SCHOTT AG Lighting and Imaging™ 158402
Product Code. 15733032
  • 166.00 € / 1 unit
Estimated Shipment: 19-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Positions the Combi light guide (Schott part 170 402) on a 32 mm diameter column.
Specifications
Specifications
Adapter
32 mm
SCHOTT Combi Lightguide (170 402)
1.3 in.
Stereo Microscopy Illumination
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SCHOTT AG Lighting and Imaging™ Column Adapter for Combi Light Guide > Column Adapter; For Use With: SCHOTT Combi Light Guide (170 402)