Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05115503
 View more versions of this product
Product Code. 10509581
  • 71.60 € / Each
Explore more special offers
Description
Description
  • Columns acc. to Hempel
  • With removable glass jacket
  • For packings like Raschig rings or glass beads
  • With NS socket and NS cone
  • With removable glass jacket
  • Made of DURAN™ tubing
Specifications
Specifications
500 mm
Cone and Socket NS 24/29