Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
47.17 € - 92.01 €
Product Code | Brand | Length (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
10141761
|
Lenz Laborglasintrumente
05115304 |
300 mm | Cone and Socket NS 29/32 |
64.02 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10020501
|
Lenz Laborglasintrumente
05115803 |
800 mm | Cone and Socket NS 24/29 |
92.01 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10129310
|
Lenz Laborglasintrumente
05115301 |
300 mm | Cone and Socket NS 14/23 |
47.17 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10224581
|
Lenz Laborglasintrumente
05115303 |
300 mm | Cone and Socket NS 24/29 |
64.02 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10509581
|
Lenz Laborglasintrumente
05115503 |
500 mm | Cone and Socket NS 24/29 |
71.60 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10749031
|
Lenz Laborglasintrumente
05115804 |
800 mm | Cone and Socket NS 29/32 |
92.01 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Columns acc. to Hempel
- With removable glass jacket
- For packings like Raschig rings or glass beads
- With NS socket and NS cone
- With removable glass jacket
- Made of DURAN™ tubing