Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™ MS10-1-2
 View more versions of this product
Product Code. 11853313
  • 24.75 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Blue
Comb
1 Each
720 μL
Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit
1 Prep, 1 Marker
2 mm