Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
24.75 € - 32.70 €
Specifications
Type Comb
Quantity 1 Each
Thickness (Metric) 2 mm
View More Specs
Products 9
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
11823313
View Documents
Fisherbrand™
MS10-10MC-2
10 54 μL
32.70 €
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
11853313
View Documents
Fisherbrand™
MS10-1-2
1 Prep, 1 Marker 720 μL
24.75 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11893313
View Documents
Fisherbrand™
MS10-12-2
12 45 μL
24.75 €
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
11833323
View Documents
Fisherbrand™
MS10-16-2
16 32 μL
24.75 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11883323
missing translation for 'viewDocuments'
Fisherbrand™
MS10-20-2
20 27 μL
24.75 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11813333
missing translation for 'viewDocuments'
Fisherbrand™
MS10-2-2
2 Prep, 2 Marker 315 μL x 8
24.75 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11853333
missing translation for 'viewDocuments'
Fisherbrand™
MS10-25-2
25 20 μL
26.80 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
11893333
missing translation for 'viewDocuments'
Fisherbrand™
MS10-4-2
4 Prep, 2 Marker 153 μL
24.75 €
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
11813353
missing translation for 'viewDocuments'
Fisherbrand™
MS10-8-2
8 81 μL
24.75 €
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
Description
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Comb
1 Each
2 mm