Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 1.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™ MS10-16-1.5
 View more versions of this product
Product Code. 11823323
  • 26.20 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Sub-Gel Midi Horizontal Gel Electrophoresis Unit
16
1.5 mm
Comb
1 Each
24 μL