Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 1.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
23.95 € - 34.80 €
Specifications
Type Comb
Quantity 1 Each
Thickness (Metric) 1.5 mm
View More Specs
Products 9
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
11813313
View Documents
Fisherbrand™
MS10-10MC-1.5
10 41 μL
23.95 €
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
11843313
View Documents
Fisherbrand™
MS10-1-1.5
1 Prep, 1 Marker 540 μL
23.95 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11883313
View Documents
Fisherbrand™
MS10-12-1.5
12 34 μL
34.15 €
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
11823323
View Documents
Fisherbrand™
MS10-16-1.5
16 24 μL
26.20 €
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
11873323
View Documents
Fisherbrand™
MS10-20-1.5
20 20 μL
34.80 €
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
11803333
View Documents
Fisherbrand™
MS10-2-1.5
2 Prep, 2 Marker 236 μL
23.95 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11843333
View Documents
Fisherbrand™
MS10-25-1.5
25 15 μL
23.95 €
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
11883333
View Documents
Fisherbrand™
MS10-4-1.5
4 Prep, 2 Marker 115 μL
23.95 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11823343
View Documents
Fisherbrand™
MS10-8-1.5
8 61 μL
23.95 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
Description
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Comb
1 Each
1.5 mm