missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for MultiSUB Maxi Gel Electrophoresis Unit, 1mm Thick

For use in preparatory electrophoresis and high-throughput screening.

Brand:  Fisherbrand™ MS20-36-1

 View more versions of this product

Product Code. 11823453

  • 34.20 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Recommended for high resolution gels and publication quality data
  • Supplied as standard
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications

Specifications

MultiSUB Maxi-Gel Electrophoresis Unit
36
1 mm
Comb
1 Each
14 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers