Learn More
Fisherbrand™ Combs for MultiSUB Maxi Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
32.50 € - 48.35 €
Specifications
For Use With (Equipment) | MultiSUB Maxi-Gel Electrophoresis Unit |
---|---|
Type | Comb |
Quantity | 1 Each |
Thickness (Metric) | 1 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11853423
|
Fisherbrand™
MS20-10-1 |
10 | 72 μL |
32.50 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||||
11883423
|
Fisherbrand™
MS20-1-1 |
1 Prep, 1 Marker | 675 μL |
32.50 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||
11823433
|
Fisherbrand™
MS20-16-1 |
16 | 41 μL |
32.50 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||
11873433
|
Fisherbrand™
MS20-20MC-1 |
20 | 27 μL |
45.50 €
Each |
In Stock
Log in to see stock. |
||||
11803443
|
Fisherbrand™
MS20-2-1 |
2 Prep, 2 Marker | 315 μL |
32.50 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||
11843443
|
Fisherbrand™
MS20-25-1 |
25 | 21 μL |
46.00 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||
11883443
|
Fisherbrand™
MS20-30-1 |
30 | 17 μL |
33.20 €
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
||||
11823453
|
Fisherbrand™
MS20-36-1 |
36 | 14 μL |
34.20 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||
11873453
|
Fisherbrand™
MS20-40MCSS-1 |
40 | 11 μL |
48.35 €
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
||||
11803463
|
Fisherbrand™
MS20-4-1 |
32.50 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||||
11843463
|
Fisherbrand™
MS20-50-1 |
50 | 10 μL |
33.20 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||
Description
- Recommended for high resolution gels and publication quality data
- Supplied as standard
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Maxi-Gel Electrophoresis Unit | |
Comb | |
1 Each | |
1 mm |