Learn More
Fisherbrand™ Combs for MultiSUB Maxi Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™ MS20-1-1
 View more versions of this product
Product Code. 11883423
  • 32.50 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Recommended for high resolution gels and publication quality data
  • Supplied as standard
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
Specifications
White
Comb
1 Each
675 μL
MultiSUB Maxi-Gel Electrophoresis Unit
1 Prep, 1 Marker
1 mm