missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for SVG-SYS Vari-gel MINI Horizontal Gel Electrophoresis Unit
For use with SVG-SYS Vari-gel MINI Horizontal Gel Electrophoresis Unit
Brand: Fisherbrand™ SVGC1512
This item is not returnable.
View return policy
Description
- Color coded
- Available in 1, 1.5 and 2mm thickness
Specifications
SVG-SYS Vari-Gel Mini Horizontal Gel Electrophoresis Unit | |
12 | |
22 μL |
Comb | |
1.5 mm | |
3 mm |