missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for SVG-SYS Vari-gel MINI Horizontal Gel Electrophoresis Unit

For use with SVG-SYS Vari-gel MINI Horizontal Gel Electrophoresis Unit

77.00 € - 93.85 €

Specifications

Type Comb
View More Specs

Products 7
Product Code Brand Width (Metric) Teeth Volume (Metric) Sample No. of Wells Thickness (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Teeth Volume (Metric) Sample No. of Wells Thickness (Metric) Price Quantity & Availability  
12960761
View Documents Promotion Details
Fisherbrand™
SVG-C1.5-15
2 mm 15 μL 15 1.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11746195
View Documents Promotion Details
Fisherbrand™
SVGC112
3 mm 15 μL 12 1 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12389808
View Documents Promotion Details
Fisherbrand™
SVG-C1-15
2 mm 10 μL 15 1 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11730996
View Documents Promotion Details
Fisherbrand™
SVGC212
3 mm 30 μL 12 2 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11720996
View Documents Promotion Details
Fisherbrand™
SVGC28
5.5 mm 54 μL 8 2 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11756195
View Documents Promotion Details
Fisherbrand™
SVGC1512
3 mm 22 μL 12 1.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12547846
View Documents Promotion Details
Fisherbrand™
SVGC158
5.5 mm 40 μL 8 1.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Color coded
  • Available in 1, 1.5 and 2mm thickness
Specifications

Specifications

Comb
Videos
SDS
Documents

Documents

Product Certifications