missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for SVG-SYS Vari-gel MINI Horizontal Gel Electrophoresis Unit

For use with SVG-SYS Vari-gel MINI Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™ SVGC158

 View more versions of this product

Product Code. 12547846

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Color coded
  • Available in 1, 1.5 and 2mm thickness
Specifications

Specifications

Comb
1.5 mm
5.5 mm
8
40 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers