missing translation for 'onlineSavingsMsg'
Learn More

3M™ Forehead Comfort Bands for use with 3M™ Versaflo™ S-Series Headcovers and Hoods

Designed to create a more comfortable fit for head covers and hoods, the 3M™ Forehead Comfort Band attaches to the head suspension headband to cushion the user's forehead. Available in various sizes.

Brand:  3M™ 7000044309

13.88 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15729027

  • 17.00 € / Pack of 5
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Product Type: Forehead Comfort Band
  • Description: Standard Format, CE-Approved
  • Color: Grey
  • For Use With (Equipment): 3M™ Versaflo™ S-Series Headcovers and Hoods
  • For Use With (Application): Respiratory Protection
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard, CE-Approved
Reusable
Small
Versaflo S-Series Headcovers and Hoods
20 Pk.
Gray
Comfort Band
Respiratory Protection
S-957S
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers