missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Compact Bucket System
BIOPHARMAGREENER_CHOICE

Contec Compact Bucket System is an all-in-one solution to make cleaning and disinfecting easier and more efficient

1460.00 € - 3035.00 €

Specifications

Disposable Reusable
Quantity 1 Ea.
View More Specs

Products 5
Product Code Brand Material Includes Product Type Price Quantity & Availability  
Product Code Brand Material Includes Product Type Price Quantity & Availability  
16348707
BIOPHARMA
View Documents
Contec™
2940P3S
Stainless Steel No Debris Screens Standard Handle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16330092
GREENER_CHOICE
View Documents
Contec™
2940U3B
Polypropylene Utility Handle
2060.00 €
Each
Estimated Shipment: 12-03-2025
to see stock.
Add to basket
 
16320092
BIOPHARMA
View Documents
Contec™
2940P3B
Polypropylene Standard Handle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16416169
BIOPHARMAGREENER_CHOICE
View Documents
Contec™
2940U2B
Polypropylene Utility Handle
1715.00 €
Each
Estimated Shipment: 12-03-2025
to see stock.
Add to basket
 
15911841
BIOPHARMAGREENER_CHOICE
View Documents
Contec™
2940P2B
Polypropylene Standard Handle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Designed for use with Contec mops such as the VertiKlean™ MAX™, VertiKlean™ MAX™ Sealed Edge, EasyCurve™ and QuickTask™ Mopping Systems
  • Stainless steel cart is available in multiple configurations to meet the unique needs of the environment
  • Available with either two or three, autoclavable polypropylene buckets, or with three stainless steel buckets, and a choice of a standard or a utility cart handle
  • Utility handle allows two small buckets (6 L) or one large bucket (15 L) to be positioned on the handle providing additional storage for extra supplies such as clean mop heads, solution spray bottles, wipes, and other items as needed
  • Cart frame has optional mop head removal tool (model no. 2639) on the front for easy, hands-free removal of a mop head
  • Two mop holders with handle clasps on either side of the cart allow for convenient mop storage during transport and storage
  • Polypropylene buckets come with a stainless steel debris screen positioned in the base of the clean and rinse buckets with a three bucket system and in the clean bucket for a two bucket system, preventing the mop from touching the bottom of the bucket where unwanted contaminants settle
Specifications

Specifications

Reusable
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications