missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Complete Assemblies Soxhlet Extractor

Quickfit™ Complete Assemblies Soxhlet Extractor includes all the glass components necessary to perform a solid-liquid extraction, including the extractor, condenser and evaporating flask.

Brand:  Quickfit™ 200RASX

Product Code. 12468786

  • 379.00 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes EX5/83 Extractor, CX7/08/SC condenser, FR500/3S Flask
  • Each assembly comprises the basic components for solid-liquid extraction : Flask/Soxhlet extractor/Condenser/Associated lids and clips
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Soxhlet Extractor
200 mL
Solid-liquid extraction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Quickfit™ Complete Assemblies Soxhlet Extractor > Capacity: 200mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.