Learn More
Acetone, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical A/0560/PC24
100.17 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description

Specifications
67-64-1 | |
10 L | |
0.32 mPaS at 20°C | |
C3H6O | |
Liquid | |
7 | |
56°C |
Plastic container | |
247mbar at 20°C | |
2 | |
8765 | |
Colorless | |
-95°C | |
58.08g/mol |
Safety and Handling
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Acetone
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527023 | Certificate of Analysis | 14/02/2025 | A/0560/21Z,A/0560/PC21,A/0560/PC24,A/0560/21,A/0560/15,A/0560/27,A/0560/25,A/0560/08,A/0560/PC21X,A/0560/17,A/0560/21S,A/0560/PC25,A/0560/PC17,A/0560/21X,A/0560/PB17,A/0560/FP21 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.