missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Concanavalin A (Con A) Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.

Brand:  Invitrogen™ C56127

169.40 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 17897378

  • 242.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Immunocytochemistry, Immunohistochemistry, Immunofluorescence
Canavlia ensiformis
α-man, α-glc
Native
Store in freezer (-5°C to -30°C) and protect from light.
Classic Dyes
Lectins
Concanavalin A (Con A)
Near-infrared
Imaging Quality
1 mg x 5 vials
Alexa Fluor Plus 750
C. ensiformis
None
750/777 nm
Room Temperature
Lyophilized
Imaging
Alexa Fluor™
Lectin
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZH402314 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog NumberC56127
Lot NumberZE399066 Certificate TypeCertificate of Analysis Date29/05/2024 Catalog NumberC56127
Lot NumberZC397103 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberC56127
Lot NumberYD373866 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog NumberC56127
Lot NumberYD374671 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog NumberC56127
Special Offers

Special Offers

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Invitrogen™ Concanavalin A (Con A) Conjugates > Alexa Fluor™ Plus 750

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt