Learn More
Lenz Laborglasintrumente Condensers Acc. to Davies
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05215303
 View more versions of this product
Product Code. 10050381
  • 155.00 € / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Davies condensers for condensing vapours
  • With NS socket and NS cone
  • With double sized cooling area
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Specifications
250 mm
NS 24/29
Borosilicate Glass (3.3)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Condensers Acc. to Davies > NS 24/29