Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05210704
 View more versions of this product
Product Code. 10171901
  • 130.00 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Liebig condenser acc. to DIN 12576 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Specifications
750 mm
NS 29/32
Borosilicate Glass (3.3)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives > 750 mm