Learn More
Lenz Laborglasintrumente Cones with Drip Tip
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100414
 View more versions of this product
Product Code. 10500132
  • 90.00 € / Pack of 10
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12249 form F
  • Conical ground cones
  • Made of DURAN™ tubing
Specifications
Specifications
NS 14/23
1.5 mm
13 mm
Borosilicate Glass (3.3)