Learn More
Lenz Laborglasintrumente Cones with Drip Tip
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
90.00 € - 107.00 €
Specifications
Material Borosilicate Glass (3.3)
View More Specs
Products 4
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
10284381
View Documents
Lenz Laborglasintrumente
01100424
NS 24/29
107.00 €
Pack of 10
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
10408792
View Documents
Lenz Laborglasintrumente
01100419
NS 19/26
105.00 €
Pack of 10
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
10418982
View Documents
Lenz Laborglasintrumente
01100429
NS 29/32
107.00 €
Pack of 10
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
10500132
View Documents
Lenz Laborglasintrumente
01100414
NS 14/23
90.00 €
Pack of 10
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12249 form F
  • Conical ground cones
  • Made of DURAN™ tubing
Specifications
Specifications
Borosilicate Glass (3.3)