missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Conical Dropping Funnels with Glass Plug
Made of borosilicate glass. Lenz Laborglasinstrumente™ Conical Dropping Funnels with Glass Plug is used to manage the slow transfer of fluids with the featured stopcock to control the flow rate.
73.00 € - 84.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11806214
|
Lenz Laborglasinstrumente™
04453702 |
100 mL |
73.00 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
11866114
|
Lenz Laborglasinstrumente™
04454904 |
250 mL |
84.00 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
Description
- Screw thread retaining device
- With inscription label, without graduation
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
Borosilicate Glass | |
Dropping Funnel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Conical Dropping Funnels with Glass Plug