missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Conical Dropping Funnels with Glass Plug

Made of borosilicate glass. Lenz Laborglasinstrumente™ Conical Dropping Funnels with Glass Plug is used to manage the slow transfer of fluids with the featured stopcock to control the flow rate.

Brand:  Lenz Laborglasinstrumente™ 04453702

 View more versions of this product

Product Code. 11806214

  • 73.00 € / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Screw thread retaining device
  • With inscription label, without graduation
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications

Specifications

100 mL
Borosilicate Glass
2.5 mm
Dropping Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasinstrumente™ Conical Dropping Funnels with Glass Plug > Capacity: 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.