Learn More
JG Finneran Associates™ Conical Inserts
Use these inserts for a variety of applications. JG Finneran™ Conical Inserts feature a polymer spring which acts as a shock absorber and protects against breakage if the needle bottoms out.
Brand:  JG Finneran Associates™ 4025BS-629
 View more versions of this product
Product Code. 12327019
  • 215.00 € / Pack of 100
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available preassembled with a polymer bottom spring or patented Top Spring™
  • Polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out
  • Conical design permits complete sample removal
Specifications
Specifications
Glass
Bottom Spring
100 Pk.
250 μL
Big mouth, limited volume, precision formed mandrel