Learn More
JG Finneran Associates™ Conical Inserts
Use these inserts for a variety of applications. JG Finneran™ Conical Inserts feature a polymer spring which acts as a shock absorber and protects against breakage if the needle bottoms out.
23.75 € - 215.00 €
Specifications
Quantity 100 Pk.
View More Specs
Products 6
Product Code Brand Volume (Metric) Style Material Includes Price Quantity & Availability  
Product Code Brand Volume (Metric) Style Material Includes Price Quantity & Availability  
12327019
View Documents
JG Finneran Associates™
4025BS-629
250 μL Big mouth, limited volume, precision formed mandrel Glass Bottom Spring
215.00 €
Pack of 100
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
12337019
View Documents
JG Finneran Associates™
4025PBS-631
250 μL Big mouth, limited volume, precision formed interior Polypropylene Bottom Spring
51.80 €
Pack of 100
In Stock
to see stock.
Add to basket
12377009
View Documents
JG Finneran Associates™
401PBS-530
100 μL Limited volume, precision formed interior Polypropylene Polymer Bottom Spring
31.70 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10537352
View Documents
JG Finneran Associates™
4005BS-625
50 μL Big mouth, limited volume, precision formed mandrel Glass Bottom Spring
75.15 €
Pack of 100
Estimated Shipment: 20-12-2024
to see stock.
Add to basket
10340571
View Documents
JG Finneran Associates™
401TS-529
100 μL Limited volume, precision formed mandrel Glass Top Spring
122.00 €
Pack of 100
Estimated Shipment: 20-12-2024
to see stock.
Add to basket
10045510
View Documents
JG Finneran Associates™
4025PF-631
250 μL Big mouth, limited volume, precision formed interior Polypropylene Plastic Flange for Step Vial This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Available preassembled with a polymer bottom spring or patented Top Spring™
  • Polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out
  • Conical design permits complete sample removal
Specifications
Specifications
100 Pk.