Learn More
JG Finneran Associates™ Conical Inserts
Use these inserts for a variety of applications. JG Finneran™ Conical Inserts feature a polymer spring which acts as a shock absorber and protects against breakage if the needle bottoms out.
Brand: JG Finneran Associates™ 4025PBS-631
Description
- Available preassembled with a polymer bottom spring or patented Top Spring™
- Polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out
- Conical design permits complete sample removal
Specifications
Polypropylene | |
Bottom Spring | |
100 Pk. |
250 μL | |
Big mouth, limited volume, precision formed interior |