missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Amplitude™ Delta™ Specialty Cleanroom Wipes, ISO Class 5
BIOPHARMAGREENER_CHOICE

Clean delicate optical products such as lenses and mirrors. Contec™ Amplitude™ Delta™ Specialty Cleanroom Wipes, ISO Class 5 are compatible with common cleaning and disinfecting solutions. Synthetic fiber build material is nonabrasive with low levels of particles and extractables.

Brand:  Contec™ AMDE0001

466.99 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 16496820

  • 656.00 € / Pack of 3600
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Extremely soft and clean
  • Low levels of particles and extractables
  • Nonabrasive texture
  • Use for cleaning delicate products
  • Wipe down products prior to introduction to cleanroom
  • Made of 100% polyester hydroentangled nonwoven for exceptional cleanliness, strength and durability
  • Ideal for use with solvents
  • Double packed to preserve product integrity

Order Info

Basis weight, 68g/m2.

Specifications

Specifications

Cleanroom Wipe
23 cm
23 cm
Common cleaning and disinfecting solutions
White
Cleanroom
9 in.
9 in.
Synthetic Fiber
ISO Class 5
9 x 9 in. (22.86 x 22.86 cm)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.