missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Amplitude™ Lambda Wipes
BIOPHARMAGREENER_CHOICE

Composed of hydraulically entangled blend of 58% cellulose and 42% polyester wipe. Contec™ Amplitude™ Lambda Wipes are strong and durable even when wet.

Brand:  Contec™ AMLA0001

180.99 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15616986

  • 254.00 € / Pack of 3600
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Excellent all-purpose wipes
  • Nonwoven strong and durable hydroentangled fabric
  • Low levels of particles and residues
  • Outstanding sorbency
Specifications

Specifications

Cleanroom Wipe
23 cm
23 cm
9 x 9 in. (22.86 x 22.86 cm)
Non-sterile
9 in.
9 in.
Polyester Cellulose
General purpose cleaning in biotech, pharmaceutical, semiconductor, electronics, medical devices, and printed circuit board industries
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.