Learn More
Contec™ Amplitude™ Lambda Wipes
Composed of hydraulically entangled blend of 58% cellulose and 42% polyester wipe. Contec™ Amplitude™ Lambda Wipes are strong and durable even when wet.
242.00 €
Specifications
Material | Polyester Cellulose |
---|---|
For Use With (Application) | General purpose cleaning in biotech, pharmaceutical, semiconductor, electronics, medical devices, and printed circuit board industries |
Description
- Excellent all-purpose wipes
- Nonwoven strong and durable hydroentangled fabric
- Low levels of particles and residues
- Outstanding sorbency
Specifications
Polyester Cellulose | |
General purpose cleaning in biotech, pharmaceutical, semiconductor, electronics, medical devices, and printed circuit board industries |