missing translation for 'onlineSavingsMsg'
Learn More

Corning™ XT Starter Ice-free Coolers
BIOPHARMA

Bench top cooling systems that provide sample cooling or freezing without ice, electricity, or batteries.

Brand:  Corning™ 432015

69.26 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15592781

  • 74.40 € / Each
Estimated Shipment: 12-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Open-platform cooler that accommodates most Corning CoolRack™ and Corning CoolSink™ modules
  • Low profile and small footprint make it ideal for use in the hood
  • Keeps samples cold (0.5°C to 4.0°C) for up to four hours
  • 1°C to 8°C temperature indicator provides visual assurance of proper temperature.
  • System may also be used with dry ice to create a compact, portable, snap-freezing workstation for bacteria, viruses, or proteins
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CoolRack and CoolSink Modules
Purple
XT Starter Holder Only
16.2 x 12.1 x 6.1 cm
Ice-free Cooler
Sample cooling or freezing without ice, electricity, or batteries
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number26218022 Certificate TypeCertificate of Compliance Date16/05/2024 Catalog Number432015
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ XT Starter Ice-free Coolers > XT Starter Holder only

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.