missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolBox™ XT Systems
BIOPHARMA

Provide sample cooling or freezing without ice, electricity, or batteries.

Brand:  Corning™ 432025

 View more versions of this product

Product Code. 15502801

  • 952.00 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning CoolBox 2XT systems are bench top cooling workstations that accomodate two CoolRacks and provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.
  • Includes two (2) XT Cooling Cores for 0.5° to 4°C cooling
  • Accommodates two (2) sample modules
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Benchtop cooling system
    Cool Box
    30 Place
    CoolBox 2XT
    Purple
    Enables Cooling for 16 Hours, Freezing for 8 Hours
    Holder and Two Cooling Cores
    5 kg
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers