Learn More
Corning™ CoolRack™ XT PCR Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Brand: Corning™ 432053
This item is not returnable.
View return policy
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.Specifications
Capacity: 96-well PCR microplate, 12 stripwells or microcentrifuge tubes, 1 SBS-compatible | |
PCR96 | |
PCR Tube Module |