missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CoolRack™ XT PCR Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
362.00 € - 411.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15582821
|
Corning™
432053 |
Capacity: 96-well PCR microplate, 12 stripwells or microcentrifuge tubes, 1 SBS-compatible |
371.00 €
Each |
In Stock
Log in to see stock. |
|||||
15592821
|
Corning™
432054 |
Cool rack for 1.5 to 2 mL tubes; holds PCR and MCT tubes |
362.00 €
Each |
In Stock
Log in to see stock. |
|||||
15502831
|
Corning™
432055 |
Capacity: 384-well PCR microplate, SBS-compatible |
411.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||||