missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ XT PCR Tube Modules
BIOPHARMA

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Brand:  Corning™ 432054

343.90 EUR valid until 2024-10-31
Use promo code "21661" to get your promotional price.


 View more versions of this product

Product Code. 15592821

  • 362.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
  • CoolRack modules ensure +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible.
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Cool rack for 1.5 to 2 mL tubes; holds PCR and MCT tubes
    M_PCR
    PCR Tube Module
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers