missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) ethoxide, Thermo Scientific™

Product Code. 11333507
Click to view available options
Quantity:
0.5 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2850-65-9
Molecular Formula C4H10CuO2
Molecular Weight (g/mol) 153.668
MDL Number MFCD00015650
InChI Key CRCKGIUJMFFISH-UHFFFAOYSA-N
Synonym copper ii ethoxide, copper diethoxide, ethanol, copper 2 salt, copper ethoxide, cupric ethoxide, acmc-20aldp, copper 2+ bis ethoxide, copper 2+ ion bis ethoxide
PubChem CID 14419876
IUPAC Name copper;ethanolate
SMILES CC[O-].CC[O-].[Cu+2]

Specifications

Melting Point ∼120°C (decomposition)
Color Green
Odor Alcohol-like
Linear Formula Cu(OC2H5)2
Quantity 25 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water and decomposes.
Formula Weight 153.67
Physical Form Powder
Chemical Name or Material Copper(II) ethoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'