Learn More
Copper(II) fluoride, 98%, anhydrous, Thermo Scientific Chemicals
144.00 € - 554.00 €
Chemical Identifiers
CAS | 7789-19-7 |
---|---|
Molecular Formula | CuF2 |
Molecular Weight (g/mol) | 101.54 |
Synonym | Cupric fluoride |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11363992
|
Thermo Scientific Acros
203370250 |
25 g | Plastic bottle |
144.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11934171
|
Thermo Scientific Acros
203371000 |
100 g | Plastic bottle |
554.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-19-7 | |
CuF2 | |
101.54 | |
Cupric fluoride |
Specifications
950.0°C | |
Blue-Gray to Gray | |
97.5% min | |
CuF2 | |
Cupric fluoride | |
101.54 | |
98% | |
4.23 | |
Copper(II) fluoride |
7789-19-7 | |
Powder | |
CuF2 | |
15,2628 | |
Solubility in water: 47g/L (20°C). Other solubilities: hydrolyzes in hot water (forming oxyfluoride), soluble in acids and alcohol | |
101.54 | |
0.5% max. | |
4.2300g/mL |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with acids liberates very toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-147-3
- Copper(II) fluoride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only