missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) sulfate, 98+%, for analysis, anhydrous, Thermo Scientific Chemicals Product Code.: 11994231

Copper(II) sulfate, 98+%, for analysis, anhydrous, Thermo Scientific Chemicals

Product Code. 11994231
2.5 kg, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11994231

Brand: Thermo Scientific Acros 197710025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.6
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagleShow MoreShow Less
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Melting Point 200.0°C
Color Gray to Green
Physical Form Fine Powder
Quantity 2.5 kg
Assay Percent Range 98% min. (Iodometry)
Linear Formula CuSO4
Fieser 01,164; 02,89; 05,162; 06,141; 08,125; 10,107; 16,283
Merck Index 15,2643
Solubility Information Solubility in water: 203g/L (20°C).
Formula Weight 159.6
Percent Purity ≥98%
Grade Analytical
Loss on Drying 1% (300°C) max.
Packaging Glass bottle
Chemical Name or Material Copper(II) sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465325 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number197710025,197715000,197711000
Lot NumberA0457322 Certificate TypeCertificate of Analysis Date06/11/2023 Catalog Number197710025,197715000,197711000
Lot NumberA0420974 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog Number197710025,197715000,197711000
Lot NumberA0401317 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number197710025,197715000,197711000
Lot NumberA0441840 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog Number197710025,197715000,197711000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) sulfate, 98+%, for analysis, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.