missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Sulfate, Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
250 g
1 kg
6 x 250 g
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Physical Form | Anhydrous |
Quantity | 250 g |
Linear Formula | CuSO4 |
UN Number | 3077 |
Formula Weight | 159.61g/mol |
Percent Purity | 99 to 100.5% (based on anhydrous substance) |
Grade | puriss. |
Vapor Pressure | 7.3mmHg (25°C) |
Density | 3.603g/mL at 25°C |
Chemical Name or Material | Copper(Ii) Sulfate, Meets analytical specification of Ph. Eur.BP, USP |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
O2000 | Certificate of Analysis | 16/12/2024 | 12852-250G |
A0430143 | Certificate of Analysis | 16/11/2023 | 12852-1KG,128520250,128521000,12852-6X1KG,12852-6X250G,12852-250G |
I3530 | Certificate of Analysis | 22/06/2023 | 12852-250G |
A0404624 | Certificate of Analysis | 13/12/2022 | 12852-1KG,128520250,128521000,12852-6X1KG,12852-6X250G,12852-250G |
A0444906 | Certificate of Analysis | 08/08/2022 | 12852-1KG,128520250,128521000,12852-6X1KG,12852-6X250G,12852-250G |
Safety and Handling


Product Identifier
- Copper(II) sulfate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Copper sulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Copper(Ii) Sulfate, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction