missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 100L 3D Tank Liner

Gusseted 100L tank liner eliminates cross contamination and the need to clean and validate tanks

Brand:  Corning™ 91-300-25

Product Code. 17293584

  • 77.50 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Tank liners are priced and sold individually

  • Extends reusable tank life
  • Each bag is irradiated and individually packed to ensure sterility

Applications:

Biopharmaceutical

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 L
0.0264 gal.
Gusseted 3D Tank Liner
LDPE
Ideal for Biopharmaceutical Application
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ 100L 3D Tank Liner > 3D Tank Liner, 100L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.